3'-Brom-4'-methylacetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3'-Bromo-4'-methylacetophenone, 1-(3-Bromo-4-methylphenyl)ethanone
M-Clarity™ Qualitätsstufe = MQ100
579734-5G
SIAL579734-5GEA
81.2
EUR
SIAL579734-5G
3'-Brom-4'-methylacetophenon, Sigma-Aldrich®
3'-Brom-4'-methylacetophenon
Formel:
C₉H₉BrO Molecular Weight: 213,0700 g/mol |
MDL:
MFCD02683854 CAS-Nummer: 40180-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...