2-Brom-4,6-difluoranisol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

579912-1G
SIAL579912-1GEA 36.5 EUR
SIAL579912-1G
2-Brom-4,6-difluoranisol, Sigma-Aldrich®
2-Brom-4,6-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00070754
CAS-Nummer: 202865-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR