4-Nitro-2-(trifluormethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

579955-5G
SIAL579955-5GEA 67.6 EUR
SIAL579955-5G
4-Nitro-2-(trifluormethyl)benzonitril, Sigma-Aldrich®
4-Nitro-2-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
Molecular Weight: 216,12 g/mol
MDL: MFCD01075729
CAS-Nummer: 320-47-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR