4-Chlorbenzoylacetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Chlorphenyl)-3-oxopropionsäurenitril, 3-(4-Chlorphenyl)-3-ketopropionsäurenitril

M-Clarity™ Qualitätsstufe = MQ100

586374-5G
SIAL586374-5GEA 36.3 EUR
SIAL586374-5G
4-Chlorbenzoylacetonitril, Sigma-Aldrich®
4-Chlorbenzoylacetonitril
Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
MDL: MFCD00051625
CAS-Nummer: 4640-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR