6-Fluor-2-methyl-4-chinolinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluoro-4-hydroxy-2-methylquinoline, 6-Fluoro-2-methylquinolin-4-ol

M-Clarity™ Qualitätsstufe = MQ100

586501-1G
SIAL586501-1GEA 78 EUR
SIAL586501-1G
6-Fluor-2-methyl-4-chinolinol, Sigma-Aldrich®
6-Fluor-2-methyl-4-chinolinol
Formel: C₁₀H₈FNO
Molecular Weight: 177,18 g/mol
MDL: MFCD00041442
CAS-Nummer: 15912-68-2
EINECS: 240-054-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR