2-Furoylacetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Furan-2-yl)-3-oxopropanenitrile, 3-(Fur-2-yl)-3-oxopropanenitrile

M-Clarity™ Qualitätsstufe = MQ100

586528-5G
SIAL586528-5GEA 50.5 EUR
SIAL586528-5G
2-Furoylacetonitril, Sigma-Aldrich®
2-Furoylacetonitril
Formel: C₇H₅NO₂
Molecular Weight: 135,12 g/mol
MDL: MFCD00052210
CAS-Nummer: 31909-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR