6-Bromoxindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

586595-500MG
SIAL586595-500MGEA 125 EUR
SIAL586595-500MG
6-Bromoxindol, Sigma-Aldrich®
6-Bromoxindol
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
MDL: MFCD02179605
CAS-Nummer: 99365-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR