3-Chlor-2-thenoesäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Chlorthiophen-2-carbonsäure, 3-Chlor-2-thiophencarbonsäure
M-Clarity™ Qualitätsstufe = MQ100
586994-5G
SIAL586994-5GEA
107
EUR
SIAL586994-5G
3-Chlor-2-thenoesäure, Sigma-Aldrich®
3-Chlor-2-thenoesäure
Formel:
C₅H₃ClO₂S Molecular Weight: 162,6 g/mol Schmelzpunkt: 187…188 °C |
MDL:
MFCD00043888 CAS-Nummer: 59337-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...