5-Brom-2-hydroxypyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromopyrimidin-2(1H)-one, 5-Bromo-2-pyrimidinone

M-Clarity™ Qualitätsstufe = MQ100

588172-1G
SIAL588172-1GEA 34.1 EUR
SIAL588172-1G
5-Brom-2-hydroxypyrimidin, Sigma-Aldrich®
5-Bromo-2-hydroxypyrimidine
Formel: C₄H₃BrN₂O
Molecular Weight: 174,98 g/mol
MDL: MFCD01318956
CAS-Nummer: 38353-06-9
EINECS: 253-896-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR