4-Nitro-3-(trifluormethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

590185-5G
SIAL590185-5GEA 170 EUR
SIAL590185-5G
4-Nitro-3-(trifluormethyl)benzonitril, Sigma-Aldrich®
4-Nitro-3-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
Molecular Weight: 216,12 g/mol
MDL: MFCD05664270
CAS-Nummer: 320-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR