Terbium(III)oxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

590509-10G 590509-2G
SIAL590509-10GEA 572 EUR
SIAL590509-10G SIAL590509-2G
Terbium(III)oxid, Sigma-Aldrich®
Terbium(III)oxid
Formel: O₃Tb₂
CAS-Nummer: 12036-41-8
EINECS: 234-849-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR