4,6-Dihydroxy-5-methylpyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methylpyrimidine-4,6-diol

M-Clarity™ Qualitätsstufe = MQ100

591025-5G
SIAL591025-5GEA 62.7 EUR
SIAL591025-5G
4,6-Dihydroxy-5-methylpyrimidin, Sigma-Aldrich®
4,6-Dihydroxy-5-methylpyrimidine
Formel: C₅H₆N₂O₂
Molecular Weight: 126,1133 g/mol
MDL: MFCD11506359
CAS-Nummer: 63447-38-1
EINECS: 254-941-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR