2-Iod-5-nitrotoluol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1-Iodo-2-methyl-4-nitrobenzene
M-Clarity™ Qualitätsstufe = MQ100
591033-5G
SIAL591033-5GEA
48.7
EUR
SIAL591033-5G
2-Iod-5-nitrotoluol, Sigma-Aldrich®
2-Iod-5-nitrotoluol
Formel:
C₇H₆INO₂ Molecular Weight: 263,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051517 CAS-Nummer: 5326-38-5 EINECS: 226-204-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...