2-(4-Chlorphenyl)ethylisothiocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

591378-5G
SIAL591378-5GEA 306 EUR
SIAL591378-5G
2-(4-Chlorphenyl)ethylisothiocyanat, Sigma-Aldrich®
2-(4-Chlorphenyl)ethylisothiocyanat
Formel: C₉H₈ClNS
Molecular Weight: 197,69 g/mol
MDL: MFCD00041127
CAS-Nummer: 17608-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR