3-Brom-4-chlorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

591580-5G
SIAL591580-5GEA 153 EUR
SIAL591580-5G
3-Brom-4-chlorbenzoesäure, Sigma-Aldrich®
3-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00079706
CAS-Nummer: 42860-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR