Benzhydrylisothiocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

591696-1G
SIAL591696-1GEA 44.3 EUR
SIAL591696-1G
Benzhydrylisothiocyanat, Sigma-Aldrich®
Benzhydrylisothiocyanat
Formel: C₁₄H₁₁NS
Molecular Weight: 225,31 g/mol
MDL: MFCD00046815
CAS-Nummer: 3550-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR