2-Chlorbenzimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-1H-benzimidazol

M-Clarity™ Qualitätsstufe = MQ100

592277-25G 592277-5G
SIAL592277-25GEA 89.3 EUR
SIAL592277-25G SIAL592277-5G
2-Chlorbenzimidazol, Sigma-Aldrich®
2-Chlorobenzimidazole
Formel: C₇H₅ClN₂
Molecular Weight: 152.58 g/mol
MDL: MFCD00051944
CAS-Nummer: 4857-06-1
EINECS: 225-453-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR