Bis(dimethylphosphino)methan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
592684-1G
SIAL592684-1GEA
186
EUR
SIAL592684-1G
Bis(dimethylphosphino)methan, Sigma-Aldrich®
Bis(dimethylphosphino)methan
Formel:
C₅H₁₄P₂ Molecular Weight: 136,11 g/mol |
MDL:
MFCD00058731 CAS-Nummer: 64065-08-3 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...