11-Chlor-1-undecen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

593117-25ML
SIAL593117-25MLEA 165 EUR
SIAL593117-25ML
11-Chlor-1-undecen, Sigma-Aldrich®
11-Chloro-1-undecene
Formel: C₁₁H₂₁Cl
Molecular Weight: 188,74 g/mol
MDL: MFCD00671354
CAS-Nummer: 872-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR