5-Brom-2-hydroxy-6-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromo-6-hydroxy-2-methylpyridine, 5-Bromo-6-methylpyridin-2-ol

M-Clarity™ Qualitätsstufe = MQ100

593206-1G
SIAL593206-1GEA 67.7 EUR
SIAL593206-1G
5-Brom-2-hydroxy-6-picolin, Sigma-Aldrich®
5-Brom-2-hydroxy-6-picolin
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD03427653
CAS-Nummer: 54923-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR