5-Fluor-3-methylsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Fluoro-2-hydroxy-3-methylbenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

593281-1G
SIAL593281-1GEA 202 EUR
SIAL593281-1G
5-Fluor-3-methylsalicylaldehyd, Sigma-Aldrich®
5-Fluor-3-methylsalicylaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,1384 g/mol
MDL: MFCD04039244
CAS-Nummer: 704884-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR