3-Chlor-5-fluorsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Chloro-5-fluoro-2-hydroxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

593400-1G
SIAL593400-1GEA 65.5 EUR
SIAL593400-1G
3-Chlor-5-fluorsalicylaldehyd, Sigma-Aldrich®
3-Chlor-5-fluorsalicylaldehyd
Formel: C₇H₄ClFO₂
Molecular Weight: 174,5569 g/mol
MDL: MFCD04039291
CAS-Nummer: 82128-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR