5-Brom-3-fluorsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-3-fluoro-2-hydroxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

593516-1G
SIAL593516-1GEA 94.3 EUR
SIAL593516-1G
5-Brom-3-fluorsalicylaldehyd, Sigma-Aldrich®
5-Brom-3-fluorsalicylaldehyd
Formel: C₇H₄BrFO₂
Molecular Weight: 219,0079 g/mol
MDL: MFCD03788530
CAS-Nummer: 251300-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR