6-Fluornicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluoro-5-pyridinecarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

593761-250MG
SIAL593761-250MGEA 87.5 EUR
SIAL593761-250MG
6-Fluornicotinsäure, Sigma-Aldrich®
6-Fluornicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD01859863
CAS-Nummer: 403-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR