2-Fluorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluoropyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

593885-250MG
SIAL593885-250MGEA 94.2 EUR
SIAL593885-250MG
2-Fluorisonicotinsäure, Sigma-Aldrich®
2-Fluorisonicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,10 g/mol
MDL: MFCD02181194
CAS-Nummer: 402-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR