2-Brombenzylmercaptan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: o-Bromtoluol-α-thiol

M-Clarity™ Qualitätsstufe = MQ100

593982-1G
SIAL593982-1GEA 63.7 EUR
SIAL593982-1G
2-Brombenzylmercaptan, Sigma-Aldrich®
2-Brombenzylmercaptan
Formel: C₇H₇BrS
Molecular Weight: 203,1 g/mol
MDL: MFCD04038925
CAS-Nummer: 143888-85-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR