2,6-Dichlorpyridin-N-oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dichloropyridine 1-oxide

M-Clarity™ Qualitätsstufe = MQ100

594059-5G
SIAL594059-5GEA 106 EUR
SIAL594059-5G
2,6-Dichlorpyridin-N-oxid, Sigma-Aldrich®
2,6-Dichlorpyridin-N-oxid
Formel: C₅H₃Cl₂NO
Molecular Weight: 163,99 g/mol
MDL: MFCD00130246
CAS-Nummer: 2587-00-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR