4-(Benzyloxy)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Benzyloxybenzeneboronic acid pinacol ester, 2-(4-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ Qualitätsstufe = MQ100

594148-5G
SIAL594148-5GEA 70.2 EUR
SIAL594148-5G
4-(Benzyloxy)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(Benzyloxy)phenylboronsäurepinakolester
Formel: C₁₉H₂₃BO₃
Molecular Weight: 310,19 g/mol
MDL: MFCD04974107
CAS-Nummer: 754226-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR