2-Amino-5-chlor-3-nitropyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Chlor-3-nitropyridin-2-amin, 5-Chlor-3-nitro-2-pyridylamin

M-Clarity™ Qualitätsstufe = MQ100

594202-25G
SIAL594202-25GEA 130 EUR
SIAL594202-25G
2-Amino-5-chlor-3-nitropyridin, Sigma-Aldrich®
2-Amino-5-chlor-3-nitropyridin
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173,56 g/mol
MDL: MFCD00092011
CAS-Nummer: 5409-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR