2-(Benzyloxy)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(2-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Benzyloxyphenylboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

594245-5G
SIAL594245-5GEA 334 EUR
SIAL594245-5G
2-(Benzyloxy)phenylboronsäurepinakolester, Sigma-Aldrich®
2-(Benzyloxy)phenylboronsäurepinakolester
Formel: C₁₉H₂₃BO₃
Molecular Weight: 310,1951 g/mol
MDL: MFCD03001334
CAS-Nummer: 1027757-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR