3-(Benzyloxy)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-[3-(Benzyloxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(3-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Benzyloxyphenylboronic acid, pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

594369-5G
SIAL594369-5GEA 247 EUR
SIAL594369-5G
3-(Benzyloxy)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Benzyloxy)phenylboronsäurepinakolester
Formel: C₁₉H₂₃BO₃
Molecular Weight: 310,1951 g/mol
MDL: MFCD05663839
CAS-Nummer: 765908-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR