2,2-Difluor-1,3-benzodioxol-5-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Difluorobenzo[d][1,3]dioxole-5-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ200

594423-5G
SIAL594423-5GEA 132 EUR
SIAL594423-5G
2,2-Difluor-1,3-benzodioxol-5-carbaldehyd, Sigma-Aldrich®
2,2-Difluor-1,3-benzodioxol-5-carbaldehyd
Formel: C₈H₄F₂O₃
Molecular Weight: 186.11 g/mol
MDL: MFCD00792420
CAS-Nummer: 656-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR