4-Acetoxy-3-methoxyphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

594474-5G
SIAL594474-5GEA 221 EUR
SIAL594474-5G
4-Acetoxy-3-methoxyphenylboronsäurepinakolester, Sigma-Aldrich®
4-Acetoxy-3-methoxyphenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₅
MDL: MFCD03789257
CAS-Nummer: 811841-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR