ω-Brom-2-chloracetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
594482-1G
594482-5G
SIAL594482-1GEA
62.8
EUR
SIAL594482-1G
SIAL594482-5G
ω-Brom-2-chloracetophenon, Sigma-Aldrich®
ω-Brom-2-chloracetophenon
Formel:
C₈H₆BrClO Lagertemperatur: Kühlschrank |
MDL:
MFCD00832993 CAS-Nummer: 5000-66-8 UN: 1760 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...