2-(Trifluormethyl)benzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Trifluormethyl)benzoesäureamid

M-Clarity™ Qualitätsstufe = MQ200

594512-1G
SIAL594512-1GEA 63.5 EUR
SIAL594512-1G
2-(Trifluormethyl)benzamid, Sigma-Aldrich®
2-(Trifluormethyl)benzamid
Formel: C₈H₆F₃NO
Molecular Weight: 189,14 g/mol
MDL: MFCD00014798
CAS-Nummer: 360-64-5
EINECS: 206-637-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR