4-Chlor-2-methylbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

594547-5G
SIAL594547-5GEA 29.9 EUR
SIAL594547-5G
4-Chlor-2-methylbenzonitril, Sigma-Aldrich®
4-Chlor-2-methylbenzonitril
Formel: C₈H₆ClN
Molecular Weight: 151,60 g/mol
MDL: MFCD00045603
CAS-Nummer: 50712-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR