ω-Brom-2,4-dihydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dihydroxyphenacylbromid, 2-Brom-1-(2,4-dihydroxyphenyl)ethan-1-on, 2-Brom-2',4'-dihydroxyacetophenon, α-Brom-2,4-dihydroxyacetophenon

M-Clarity™ Qualitätsstufe = MQ100

594598-1G
SIAL594598-1GEA 95 EUR
SIAL594598-1G
ω-Brom-2,4-dihydroxyacetophenon, Sigma-Aldrich®
ω-Brom-2,4-dihydroxyacetophenon
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
MDL: MFCD03789098
CAS-Nummer: 2491-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR