3-(Methoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(3-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxycarbonylphenylboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

594652-1G
SIAL594652-1GEA 33.2 EUR
SIAL594652-1G
3-(Methoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
Molecular Weight: 262,1093 g/mol
MDL: MFCD03789258
CAS-Nummer: 480425-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR