4-Fluor-o-toluonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

594660-5G
SIAL594660-5GEA 128 EUR
SIAL594660-5G
4-Fluor-o-toluonitril, Sigma-Aldrich®
4-Fluor-o-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
MDL: MFCD03095106
CAS-Nummer: 147754-12-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR