Methyl-3,4-difluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3,4-difluorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

594717-5G
SIAL594717-5GEA 45 EUR
SIAL594717-5G
Methyl-3,4-difluorbenzoat, Sigma-Aldrich®
Methyl-3,4-difluorbenzoat
Formel: C₈H₆F₂O₂
Molecular Weight: 172.13 g/mol
MDL: MFCD00276597
CAS-Nummer: 369-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR