Methyl-2,5-difluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2,5-difluorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

594822-5G
SIAL594822-5GEA 43 EUR
SIAL594822-5G
Methyl-2,5-difluorbenzoat, Sigma-Aldrich®
Methyl-2,5-difluorbenzoat
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
MDL: MFCD03093800
CAS-Nummer: 362601-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR