3-(4-Bromphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

595438-5G
SIAL595438-5GEA 70.6 EUR
SIAL595438-5G
3-(4-Bromphenyl)propionsäure, Sigma-Aldrich®
3-(4-Bromphenyl)propionsäure
Formel: C₉H₉BrO₂
MDL: MFCD01310793
CAS-Nummer: 1643-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR