4,6-Dichlor-5-methylpyrimidin, Sigma-Aldrich®

Lieferant: Merck

595446-5G
SIAL595446-5GEA 152 EUR
SIAL595446-5G
4,6-Dichlor-5-methylpyrimidin, Sigma-Aldrich®
4,6-Dichloro-5-methylpyrimidine
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
MDL: MFCD01646112
CAS-Nummer: 4316-97-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR