1-Chlor-5-(trimethylsilyl)-4-pentin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

595918-5G
SIAL595918-5GEA 34 EUR
SIAL595918-5G
1-Chlor-5-(trimethylsilyl)-4-pentin, Sigma-Aldrich®
1-Chlor-5-(trimethylsilyl)-4-pentin
Formel: C₈H₁₅ClSi
Molecular Weight: 174,75 g/mol
MDL: MFCD00671353
CAS-Nummer: 77113-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR