4-Amino-2-chlorpyrimidin-5-carbosäurenitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-2-chloro-5-pyrimidinecarbonitrile

M-Clarity™ Qualitätsstufe = MQ100

597600-1G
SIAL597600-1GEA 85 EUR
SIAL597600-1G
4-Amino-2-chlorpyrimidin-5-carbosäurenitril, Sigma-Aldrich®
4-Amino-2-chlorpyrimidin-5-carbosäurenitril
Formel: C₅H₃ClN₄
Molecular Weight: 154,56 g/mol
MDL: MFCD00052343
CAS-Nummer: 94741-69-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR