5-Acetyl-2-methoxypyridin, Sigma-Aldrich®

Lieferant: Merck

597767-5G
SIAL597767-5GEA 57.4 EUR
SIAL597767-5G
5-Acetyl-2-methoxypyridin, Sigma-Aldrich®
5-Acetyl-2-methoxypyridin
Formel: C₈H₉NO₂
MDL: MFCD05664350
CAS-Nummer: 213193-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR