4,7-Dichlorisatin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4,7-Dichlor-2,3-indolindion, 4,7-Dichlor-1H-indol-2,3-dion, 4,7-Dichlor-2,3-dioxoindolin, 4,7-Dichlorindolin-2,3-dion

M-Clarity™ Qualitätsstufe = MQ100

597813-1G
SIAL597813-1GEA 95.8 EUR
SIAL597813-1G
4,7-Dichlorisatin, Sigma-Aldrich®
4,7-Dichlorisatin
Formel: C₈H₃Cl₂NO₂
Molecular Weight: 216,02 g/mol
MDL: MFCD00047214
CAS-Nummer: 18711-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR