2-[(Trimethylsilyl)ethinyl]anisol, Sigma-Aldrich®

Lieferant: Merck

598054-5G
SIAL598054-5GEA 164 EUR
SIAL598054-5G
2-[(Trimethylsilyl)ethinyl]anisol, Sigma-Aldrich®
2-[(Trimethylsilyl)ethinyl]anisol
Formel: C₁₂H₁₆OSi
MDL: MFCD04039988
CAS-Nummer: 40230-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR