1-(2,4,6-Trichlorphenyl)-2-thioharnstoff, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,6-Trichlorphenylthioharnstoff, N-(2,4,6-Trichlorphenyl)thioharnstoff

M-Clarity™ Qualitätsstufe = MQ100

598143-1G
SIAL598143-1GEA 41.7 EUR
SIAL598143-1G
1-(2,4,6-Trichlorphenyl)-2-thioharnstoff, Sigma-Aldrich®
1-(2,4,6-Trichlorphenyl)-2-thioharnstoff
Formel: (C₆H₂Cl₃)NC(S)NH₂
Molecular Weight: 255,55 g/mol
MDL: MFCD00041153
CAS-Nummer: 31118-87-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR