n-Nonyl-β-D-maltosid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: NM

59965-1G
SIAL59965-1GEA 203 EUR
SIAL59965-1G
n-Nonyl-β-D-maltosid, Sigma-Aldrich®
n-Nonyl-β-D-maltosid
Formel: C₂₁H₄₀O₁₁
Molecular Weight: 468,54 g/mol
MDL: MFCD08062400
CAS-Nummer: 106402-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR